Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-3-Hydroxypyrrolidine hydrochloride, 98%

Catalog No. AAH6497522
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 86070-82-8 | C4H10ClNO | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86070-82-8
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD03001754
InChI Key QPMSJEFZULFYTB-UHFFFAOYNA-N
Synonym pyrrolidin-3-ol hydrochloride, 3-hydroxypyrrolidine hydrochloride, 3-pyrrolidinol hydrochloride, 3-hydroxypyrrolidine hcl, 3-pyrrolidinol hcl, 3-pyrrolidinol, hydrochloride, 3-hydroxypyrrolidinehydrochloride, s-3-hydroxypyrrolidine hcl, 3-pyrrolidinol hci, hydroxypyrrolidine hcl
PubChem CID 2769408
IUPAC Name pyrrolidin-3-ol;hydrochloride
SMILES Cl.OC1CCNC1

Specifications

Quantity 100 g
Beilstein 7029051
Formula Weight 123.58
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (±)-3-Hydroxypyrrolidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.