Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxytetrahydrofuran, 97%

Catalog No. p-3736706
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 453-20-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-20-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00005374
InChI Key XDPCNPCKDGQBAN-UHFFFAOYNA-N
Synonym 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol
PubChem CID 9960
IUPAC Name oxolan-3-ol
SMILES OC1CCOC1

Specifications

Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 1.0900g/mL
Boiling Point 181°C
Quantity 5 g
Flash Point 81°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4490 to 1.451
Assay Percent Range 96% min. (GC)
Specific Gravity 1.09
Beilstein 17, 107
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material 3-Hydroxytetrahydrofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 207-219-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.