Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indolepropanol 98.0+%, TCI America™
SDP

Catalog No. H06801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06801G 1 g
H06805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06801G Supplier TCI America Supplier No. H06801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3569-21-9
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00013437
InChI Key LYPSVQXMCZIRGP-UHFFFAOYSA-N
Synonym 3-(3-Hydroxypropyl)indole, Homotryptophol
PubChem CID 77115
IUPAC Name 3-(1H-indol-3-yl)propan-1-ol
SMILES OCCCC1=CNC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 200°C
Quantity 1 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Indolepropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.