Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indolepropionic acid, 99%

Catalog No. AC204990100
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle

CAS: 830-96-6 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-96-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005660
InChI Key GOLXRNDWAUTYKT-UHFFFAOYSA-N
Synonym 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
PubChem CID 3744
ChEBI CHEBI:43580
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)CCC1=CNC2=CC=CC=C12

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 130°C to 136°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 22, 69
Solubility Information Solubility in water: slightly soluble
Formula Weight 189.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolepropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 212-600-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.