Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Iodo-1-methyl-1H-pyrazole, 97%

Click to view available options
Quantity:
250mg
1g
92525-10-5
C4H5IN2
208.002
MFCD08435890
SITJXRWLFQGWCB-UHFFFAOYSA-N
3-iodo-1-methyl-1h-pyrazole,1-methyl-3-iodopyrazole,1h-pyrazole, 3-iodo-1-methyl,3-iodo-1-methy-1h-pyrazole,iodo-n-methylpyrazole,3-idio-1-methylpyrazole,3-iodanyl-1-methyl-pyrazole,1h-pyrazole,3-iodo-1-methyl,3 5-iodopyrazole, 1-me,3-iodo-1-methyl-1h-pyrazole
13334057
3-iodo-1-methylpyrazole
CN1C=CC(=N1)I
92525-10-5
C4H5IN2
208.002
MFCD08435890
SITJXRWLFQGWCB-UHFFFAOYSA-N
3-iodo-1-methyl-1h-pyrazole,1-methyl-3-iodopyrazole,1h-pyrazole, 3-iodo-1-methyl,3-iodo-1-methy-1h-pyrazole,iodo-n-methylpyrazole,3-idio-1-methylpyrazole,3-iodanyl-1-methyl-pyrazole,1h-pyrazole,3-iodo-1-methyl,3 5-iodopyrazole, 1-me,3-iodo-1-methyl-1h-pyrazole
13334057
3-iodo-1-methylpyrazole
CN1C=CC(=N1)I
Quantity 1g
Color Brown
Density 1.936g/mL
Refractive Index 1.5880
Sensitivity Light Sensitive
Formula Weight 208
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Iodo-1-methyl-1H-pyrazole