Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-4-methylaniline, 98%

Catalog No. AAA1148514
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 35944-64-0 | C7H8IN | 233.05 g/mol

3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35944-64-0
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00047843
InChI Key RRUDMHNAMZFNEK-UHFFFAOYSA-N
Synonym 3-iodo-p-toluidine, benzenamine, 3-iodo-4-methyl, 4-amino-2-iodotoluene, p-toluidine, 3-iodo, 3-iodo-4-methyl-aniline, benzenamine, 3-iodo-4-methyl-9ci, 3-iodo-4-methyl-phenylamine, pubchem3314, 3-iodo-4-methyl aniline, acmc-209ik4
PubChem CID 118889
IUPAC Name 3-iodo-4-methylaniline
SMILES CC1=CC=C(N)C=C1I

Specifications

Melting Point 34°C to 37°C
Quantity 25 g
UN Number UN2811
Beilstein 2353567
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 233.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-803-2
RTECSNumber XU7700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.