Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzoic acid, 98+%

Catalog No. p-7081513
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 618-51-9 | C7H5IO2 | 248.019 g/mol

3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in Chloroform and Methanol. Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-51-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002496
InChI Key KVBWBCRPWVKFQT-UHFFFAOYSA-N
Synonym m-iodobenzoic acid, benzoic acid, 3-iodo, 3-iodo-benzoic acid, benzoic acid, m-iodo, 3-iodobenzoicacid, 3-carboxyiodobenzene, 3-iodo benzoic acid, pubchem3925, acmc-209mwv, meta-carboxy iodo-benzene
PubChem CID 12060
IUPAC Name 3-iodobenzoic acid
SMILES C1=CC(=CC(=C1)I)C(=O)O

Specifications

Melting Point 186°C to 188°C
Quantity 10 g
Beilstein 971088
Sensitivity Light sensitive
Solubility Information Soluble in Chloroform and Methanol. Insoluble in water.
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-555-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.