Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodopropionic Acid 98.0+%, TCI America™
SDP

Catalog No. I007125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I007125G 25 g
I00715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I007125G Supplier TCI America Supplier No. I007125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-76-4
Molecular Formula C3H5IO2
Molecular Weight (g/mol) 199.98
MDL Number MFCD00002765
InChI Key KMRNTNDWADEIIX-UHFFFAOYSA-N
Synonym 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
PubChem CID 8856
IUPAC Name 3-iodopropanoic acid
SMILES OC(=O)CCI

Specifications

Melting Point 82°C
Quantity 25 g
UN Number 3261
Formula Weight 199.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodopropionic Acid
RTECSNumber UF4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.