Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodotoluene, 99%

Catalog No. p-7081151
Click to view available options
Quantity:
50 g
100 g
250 g

CAS: 625-95-6 | C7H7I | 218.037 g/mol

3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-95-6
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001050
InChI Key VLCPISYURGTGLP-UHFFFAOYSA-N
Synonym 3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
PubChem CID 12268
IUPAC Name 1-iodo-3-methylbenzene
SMILES CC1=CC(=CC=C1)I

Specifications

Melting Point -27°C
Density 1.698
Boiling Point 80°C to 82°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.604
Quantity 50 g
Beilstein 1903634
Sensitivity Light sensitive
Solubility Information Not miscible in water. Soluble in benzene,alcohol and ether.
Formula Weight 218.04
Percent Purity 99%
Chemical Name or Material 3-Iodotoluene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-918-5
RTECSNumber DA3440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.