Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Maleimidopropionic acid, 95%

Catalog No. aah6153603
Click to view available options
Quantity:
1 g
5 g

CAS: 7423-55-4 | C7H7NO4 | 169.14 g/mol

3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.

Notes
Moisture sensitive. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-55-4
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00043030
InChI Key IUTPJBLLJJNPAJ-UHFFFAOYSA-N
Synonym 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw
PubChem CID 573621
IUPAC Name 3-(2,5-dioxopyrrol-1-yl)propanoic acid
SMILES OC(=O)CCN1C(=O)C=CC1=O

Specifications

Melting Point 108°C to 112°C
Color White
Quantity 1 g
Beilstein 1528952
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol,dimethyl formamide,alcohol and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 169.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Maleimidopropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.