Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. M0814100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0814100G 100 g
M081425G 25 g
M0814500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0814100G Supplier TCI America Supplier No. M0814100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
MDL Number MFCD00005229
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
PubChem CID 2723802
IUPAC Name 2,3-dihydro-1H-1,2,4-triazole-3-thione
SMILES S=C1NNC=N1

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 100 g
Formula Weight 101.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Mercapto-1,2,4-triazole
EINECSNumber (5)-0606
RTECSNumber XZ5267500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.