Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1-hexanol, 96%

Catalog No. p-7026137
Click to view available options
Quantity:
5 g
25 g

CAS: 51755-83-0 | C6H14OS | 134.237 g/mol

3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51755-83-0
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.237
MDL Number MFCD00792515
InChI Key TYZFMFVWHZKYSE-UHFFFAOYSA-N
Synonym 3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
PubChem CID 521348
ChEBI CHEBI:77690
IUPAC Name 3-sulfanylhexan-1-ol
SMILES CCCC(CCO)S

Specifications

Density 0.974
Boiling Point 250°C
Flash Point >100°C (212°F)
Refractive Index 1.48
Quantity 5 g
Beilstein 1846873
Solubility Information Insoluble in water.
Formula Weight 134.24
Percent Purity 96%
Chemical Name or Material 3-Mercapto-1-hexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.