Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-2-pentanone, 97%, stab. with 0.1% Calcium carbonate

Catalog No. p-7026026
Click to view available options
Quantity:
1 g
5 g

CAS: 67633-97-0 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67633-97-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD02093290
InChI Key SZECUQRKLXRGSJ-UHFFFAOYSA-N
Synonym 3-mercapto-2-pentanone, 2-pentanone, 3-mercapto, 3-mercaptopentan-2-one, fema no. 3300, 3-mercapto-2-penanone, 2-pentanone,3-mercapto, acmc-1b6ys, 2-pentanone, 3-mercapto-7ci,9ci
PubChem CID 62237
IUPAC Name 3-sulfanylpentan-2-one
SMILES CCC(C(=O)C)S

Specifications

Density 0.988
Refractive Index 1.466
Quantity 1 g
UN Number UN3336
Sensitivity Light sensitive
Formula Weight 118.19
Percent Purity 98%
Chemical Name or Material 3-Mercapto-2-pentanone, stabilized with 0.1% Calcium carbonate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 266-799-5
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.