Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercaptohexyl Acetate 97.0+%, TCI America™
SDP

Catalog No. M249725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M249725G 25 g
M24975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M249725G Supplier TCI America Supplier No. M249725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136954-20-6
Molecular Formula C8H16O2S
Molecular Weight (g/mol) 176.274
MDL Number MFCD00792516
InChI Key JUCARGIKESIVLB-UHFFFAOYSA-N
Synonym Acetic Acid 3-Mercaptohexyl Ester
PubChem CID 518810
ChEBI CHEBI:77818
IUPAC Name 3-sulfanylhexyl acetate
SMILES CCCC(CCOC(=O)C)S

Specifications

Color Colorless
Quantity 25 g
Formula Weight 176.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Mercaptohexyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.