Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercaptophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193925 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. M24171G

Supplier: TCI America M24171G

Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 3-Mercaptophenylboronic Acid (contains varying amounts of Anhydride)
CAS 352526-01-3
Melting Point 201°C
Color White-Yellow
Molecular Formula C6H7BO2S
Quantity 1 g
MDL Number MFCD03095131
Synonym 3-Mercaptobenzeneboronic Acid
InChI Key OBCKMTLUSKYKIW-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)S)(O)O
IUPAC Name (3-sulfanylphenyl)boronic acid
Molecular Weight (g/mol) 153.99
PubChem CID 2773922
Formula Weight 153.99
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.