Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Mercaptopropyl)trimethoxysilane

Catalog No. 17561725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-561-725G 25 g
17-561-7500G 500 g
17-561-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-561-725G Supplier Sigma Aldrich Supplier No. 17561725G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-mercaptopropyl)trimethoxysilane (MPTMS) is an organosilane with a thiol group, which can be used as a self-assembled monolayer (SAM). It can be used as a conjunction reagent to immobilize a variety of nanoparticles.MPTMS can be coated with TiO2, CeO2 and La2O3 for protection against corrosion on quaternary bronze in urban atmosphere. It can also be used as a silica nanofluid (NF) to modify the surface of berea sandstone to improve the enhanced oil recovery (EOR) area in the petroleum industry. MPTMS forms composites with kaolin, that can be used as an absorbent for the removal of heavy metals from wastewater. MPTMS can form a bilayer with multiwalled carbon nanotubes (MWNTs) on gold electrodes which can be used to study the electrochemical properties of fluphenazine.

Specifications

Boiling Point 213°C to 215°C
CAS 4420-74-0
Quantity 25 g
Molecular Formula C6H16O3SSi
Linear Formula HS(CH2)3 Si(OCH3)3
MDL Number MFCD00004901
Synonym 3-(Trimethoxysilyl)-1-propanethiol
Molecular Weight (g/mol) 196.34
Percent Purity 95%
Refractive Index n20/D 1.444 (literature)
Density 1.057 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.