Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1,2-propanediol, 98%

Catalog No. p-3758336
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 623-39-2 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-39-2
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00004719
InChI Key PSJBSUHYCGQTHZ-UHFFFAOYNA-N
Synonym 3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
PubChem CID 92822
IUPAC Name 3-methoxypropane-1,2-diol
SMILES COCC(O)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Boiling Point 133°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.445
Linear Formula H3COCH2CH(OH)CH2OH
Quantity 1 g
Specific Gravity 1.12
Solubility Information Solubility in water: soluble in water
Formula Weight 106.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1, 2-propanediol
Show More Show Less
EINECSNumber 210-791-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.