Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1-propanol 98.0+%, TCI America™
SDP

Catalog No. M1475250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1475250G 250 g
M147525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1475250G Supplier TCI America Supplier No. M1475250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1589-49-7
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00039550
InChI Key JDFDHBSESGTDAL-UHFFFAOYSA-N
Synonym 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol
PubChem CID 74116
IUPAC Name 3-methoxypropan-1-ol
SMILES COCCCO

Specifications

Color Yellow
Boiling Point 153°C
Quantity 250 g
UN Number 1993
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-propanol
RTECSNumber UB7650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.