Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-methylaniline, 99+%

Catalog No. AAH5036203
Click to view available options
Quantity:
250 mg
1 g

CAS: 16452-01-0 | C8H11NO | 137.18 g/mol

3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16452-01-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00025371
InChI Key ONADZNBSLRAJFW-UHFFFAOYSA-N
Synonym o-cresidine, benzenamine, 3-methoxy-4-methyl, 3-methoxy-4-methyl aniline, ccris 4606, unii-v82scc5x3q, 5-amino-2-methylanisole, 5-amino-2-methyl-anisol, 2-methoxy-4-aminotoluene, 3-methoxy-4-methyl-aniline, v82scc5x3q
PubChem CID 27882
IUPAC Name 3-methoxy-4-methylaniline
SMILES COC1=CC(N)=CC=C1C

Specifications

Melting Point 56°C to 60°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 137.18
Percent Purity ≥99%
Chemical Name or Material 3-Methoxy-4-methylaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 240-500-8
RTECSNumber CY0888000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.