Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-5-methylphenol, 97%

Catalog No. p-7029991
Click to view available options
Quantity:
1 g
5 g

CAS: 3209-13-0 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3209-13-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00059261
InChI Key NOTCZLKDULMKBR-UHFFFAOYSA-N
Synonym 3-hydroxy-5-methoxytoluene, phenol, 3-methoxy-5-methyl, 5-methoxy-m-cresol, orcinol monomethyl ether, o-methylorcinol, 3-methoxy-5-methyl-phenol, unii-uya9w3l847, 5-methoxy-3-methylphenol, orcinyl ter, 3-methoxy-5-methylphenol
PubChem CID 76674
IUPAC Name 3-methoxy-5-methylphenol
SMILES CC1=CC(=CC(=C1)OC)O

Specifications

Melting Point 60°C to 66°C
Quantity 1 g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-716-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.