Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybenzaldehyde, 98%

Catalog No. p-7021571
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 591-31-1 | C8H8O2 | 136.15 g/mol

3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

Solubility
Slightly soluble in water. Soluble in glacial acetic acid, alcohol, chloroform, ether, carbon disulfide, methanol, aqueous solutions of alkali hydroxides.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-31-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003361
InChI Key WMPDAIZRQDCGFH-UHFFFAOYSA-N
Synonym m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
PubChem CID 11569
IUPAC Name 3-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1)C=O

Specifications

Density 1.118
Boiling Point 231°C to 233°C
Flash Point 121°C (250°F)
Odor Characteristic
Refractive Index 1.553
Quantity 25 g
Beilstein 606013
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water. Soluble in glacial acetic acid,alcohol,chloroform,ether,carbon disulfide,methanol,aqueous solutions of alkali hydroxides.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzaldehyde
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 209-712-8
RTECSNumber BZ2605000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.