Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybenzyl alcohol, 97+%

Catalog No. p-7022611
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547114 25 g
AAA1547122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1547114 Supplier Thermo Scientific Chemicals Supplier No. A1547114
Only null left
Add to Cart

CAS: 6971-51-3 | C8H10O2 | 138.166 g/mol

3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in ether, and alcohol. Insoluble in water.

Notes
Incompatible with Oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6971-51-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004637
InChI Key IIGNZLVHOZEOPV-UHFFFAOYSA-N
Synonym 3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol
PubChem CID 81437
ChEBI CHEBI:1580
IUPAC Name (3-methoxyphenyl)methanol
SMILES COC1=CC=CC(=C1)CO

Specifications

Density 1.112
Boiling Point 252°C to 254°C
Flash Point 109°C (228°F)
Refractive Index 1.544
Quantity 25 g
Beilstein 2042063
Solubility Information Soluble in ether,and alcohol. Insoluble in water.
Formula Weight 138.17
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzyl alcohol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-200-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.