Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybutyl Acetate 99.0+%, TCI America™
SDP

Catalog No. A075925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A075925ML 25 mL
A0759500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A075925ML Supplier TCI America Supplier No. A075925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4435-53-4
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD00043928
InChI Key QMYGFTJCQFEDST-UHFFFAOYNA-N
Synonym Acetic Acid 3-Methoxybutyl Ester
PubChem CID 20498
IUPAC Name 3-methoxybutyl acetate
SMILES COC(C)CCOC(C)=O

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 mL
Formula Weight 146.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybutyl Acetate
EINECSNumber (2)-0739
RTECSNumber EL4725000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.