Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxymethylindole 98.0+%, TCI America™
SDP

Catalog No. M130525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130525G 25 g
M13051G 1 g
M13055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M130525G Supplier TCI America Supplier No. M130525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78440-76-3
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00276420
InChI Key RJWLFBCRNXZXTJ-UHFFFAOYSA-N
PubChem CID 154411
IUPAC Name 3-(methoxymethyl)-1H-indole
SMILES COCC1=CNC2=CC=CC=C21

Specifications

Melting Point 98°C
Quantity 25 g
Formula Weight 161.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxymethylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.