Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methoxyphenylacetylene, 96%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is a useful aromatic compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful aromatic compound.
Solubility
Slightly soluble in water.
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
It is a useful aromatic compound.
Solubility
Slightly soluble in water.
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications
Specifications
| Density | 1.04 |
| Boiling Point | 204°C to 210°C |
| Flash Point | 76°C (169°F) |
| Refractive Index | 1.555 |
| Quantity | 5 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 132.16 |
| Percent Purity | 96% |
| Chemical Name or Material | 3-Methoxyphenylacetylene |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction