Alfa Aesar™ 3-Methoxyphenylacetylene, 96%

Manufacturer: Alfa Aesar™ H55254MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55254MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 768-70-7
    Molecular Formula C9H8O
    Formula Weight 132.16
    MDL Number MFCD00160810
    Synonym 3-Ethynylanisole; 1-Ethynyl-3-methoxybenzene
    Chemical Name or Material 3-Methoxyphenylacetylene

    Specifications

    Formula Weight 132.16
    Molecular Formula C9H8O
    Boiling Point 204° to 210°C
    Quantity 250mg
    Flash Point 76°C (169°F)
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    MDL Number MFCD00160810
    CAS 768-70-7
    Synonym 3-Ethynylanisole; 1-Ethynyl-3-methoxybenzene
    Health Hazard 1 Warning
    Chemical Name or Material 3-Methoxyphenylacetylene
    Assay 96%

    Description

    Applications
    It is a useful aromatic compound.

    Solubility
    Slightly soluble in water.

    Notes
    Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit