Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenylacetylene, 96%

Catalog No. p-7036831
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 768-70-7 | C9H8O | 132.16 g/mol

It is a useful aromatic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful aromatic compound.

Solubility
Slightly soluble in water.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-70-7
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00160810
InChI Key ZASXCTCNZKFDTP-UHFFFAOYSA-N
Synonym 3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
PubChem CID 640753
IUPAC Name 1-ethynyl-3-methoxybenzene
SMILES COC1=CC=CC(=C1)C#C

Specifications

Density 1.04
Boiling Point 204°C to 210°C
Flash Point 76°C (169°F)
Refractive Index 1.555
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 132.16
Percent Purity 96%
Chemical Name or Material 3-Methoxyphenylacetylene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.