Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxysalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. M115325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115325G 25 g
M11535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M115325G Supplier TCI America Supplier No. M115325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-22-5
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002445
InChI Key AUZQQIPZESHNMG-UHFFFAOYSA-N
Synonym 3-methoxysalicylic acid, o-vanillic acid, acide orthovanillique, benzoic acid, 2-hydroxy-3-methoxy, m-anisic acid, 2-hydroxy, 3-hydroxy-m-anisic acid, unii-3j31y0e2im, 2-hydroxy-3-methoxy-benzoic acid, 3-methoxysalicyclic acid, 2-hydroxy-3-methoxy benzoic acid
PubChem CID 70140
ChEBI CHEBI:68496
IUPAC Name 2-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 151°C
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxysalicylic Acid
RTECSNumber BZ4840000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.