Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-pentanol, 98%

Catalog No. p-7026383
Click to view available options
Quantity:
5 g
25 g

CAS: 589-35-5 | C6H14O | 102.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-35-5
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002937
InChI Key IWTBVKIGCDZRPL-UHFFFAOYSA-N
Synonym 3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
PubChem CID 11508
ChEBI CHEBI:87381
IUPAC Name 3-methylpentan-1-ol
SMILES CCC(C)CCO

Specifications

Density 0.823
Boiling Point 151°C to 152°C
Flash Point 58°C (136°F)
Refractive Index 1.418
Quantity 5 g
UN Number UN1987
Beilstein 1718979
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-pentanol
Hazard Category H226
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-644-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.