Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-penten-4-yn-3-ol 98.0+%, TCI America™
SDP

Catalog No. M131225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M131225ML 25 mL
M13125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M131225ML Supplier TCI America Supplier No. M131225ML

Chemical Identifiers

CAS 3230-69-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD01709006
InChI Key VBATUBQIYXCZPA-UHFFFAOYNA-N
Synonym 3-methyl-1-penten-4-yn-3-ol, 1-penten-4-yn-3-ol, 3-methyl, ethynyl methyl vinyl carbinol, 3-methyl-pent-4-en-1-yn-3-ol, 3-methyl pent-4-en-l-yn-3-0l, acmc-209hsd, 4-01-00-02309 beilstein handbook reference, ksc496k7h, wln: 1uu1xq1&1u1, 3-methyl-4-penten-1-yn-3-ol
PubChem CID 97815
IUPAC Name 3-methylpent-1-en-4-yn-3-ol
SMILES CC(O)(C=C)C#C

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 mL
UN Number 1987
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-penten-4-yn-3-ol
RTECSNumber SB4950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.