Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals

Catalog No. aal1152514
Click to view available options
Quantity:
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-20-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009339
InChI Key LDTAOIUHUHHCMU-UHFFFAOYSA-N
PubChem CID 12969
IUPAC Name 3-methylpent-1-ene
SMILES CCC(C)C=C

Specifications

Melting Point -154°C
Density 0.67
Boiling Point 53°C to 54°C
Flash Point −33°C (−27°F)
Odor Petrol-like
Refractive Index 1.387
Quantity 25 g
UN Number UN3295
Beilstein 1719110
Synonym 3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-pentene
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-076-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.