Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-pentene 99%, Thermo Scientific™

Catalog No. AC149980050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-20-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009339
InChI Key LDTAOIUHUHHCMU-UHFFFAOYSA-N
Synonym 3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr
PubChem CID 12969
IUPAC Name 3-methylpent-1-ene
SMILES CCC(C)C=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.6700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Linear Formula C2H5CH(CH3)CH=CH2
Quantity 5 g
Formula Weight 84.15
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-pentene, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.