Alfa Aesar™ 3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%

Manufacturer: Alfa Aesar™ A1179206

Quantity 5g

 View more versions of this product

Catalog No. AAA1179206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 707-61-9
    Molecular Formula C11H13OP
    Formula Weight 192.19
    MDL Number MFCD00014518
    Beilstein 609223
    Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide

    Specifications

    Formula Weight 192.19
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00014518
    CAS 707-61-9
    Health Hazard 1 Warning
    Melting Point 60° to 65°C
    Boiling Point 173° to 174°C (0.7mmHg)
    Beilstein 609223
    Quantity 5g
    Odor Unpleasant
    RTECS Number SZ6105100
    TSCA TSCA
    Molecular Formula C11H13OP
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide
    Assay 94%

    Description

    Applications
    3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

    Solubility
    Soluble in polar solvents.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit