Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%, Thermo Scientific Chemicals

Catalog Number AAA1116122
Click to view available options
Quantity:
100 g
500 g
2500 g
89-25-8
C10H10N2O
174.203
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2
89-25-8
C10H10N2O
174.203
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2

3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

Solubility
Soluble in water(3.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 127°C to 131°C
Density 1.12
Boiling Point 191°C (17 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 609575
Merck Index 14,6713
Solubility Information Soluble in water(3.3g/L).
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-891-0
RTECSNumber UQ9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit