Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-phenyl-3-pentanol 98.0+%, TCI America™
SDP

Catalog No. M201125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M201125G 25 g
M20115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M201125G Supplier TCI America Supplier No. M201125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10415-87-9
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00021825
InChI Key AEJRTNBCFUOSEM-UHFFFAOYSA-N
PubChem CID 61516
IUPAC Name 3-methyl-1-phenylpentan-3-ol
SMILES CCC(C)(CCC1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 130°C
Quantity 25 g
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-phenyl-3-pentanol
EINECSNumber (3)-1044
RTECSNumber SA7624000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.