Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-phenylindolin-2-one, 97%

Catalog No. AA4380806
Click to view available options
Quantity:
1 g
5 g

CAS: 23210-22-2 | C15H13NO | 223.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-22-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00798609
InChI Key DJKASMOURATDIE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenylindolin-2-one, 3-methyl-1-phenylindoline-2-one, 3-methyl-1-phenyl-2,3-dihydro-1h-indol-2-one, acmc-1ciq4, 1-phenyl-3-methylindoline-2-one, 3-methyl-1-phenyl-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one,1,3-dihydro-3-methyl-1-phenyl
PubChem CID 4990835
IUPAC Name 3-methyl-1-phenyl-3H-indol-2-one
SMILES CC1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3

Specifications

Quantity 5 g
Formula Weight 223.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Methyl-1-phenylindolin-2-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.