Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, 0.1% (w/v) in 90% (v/v) Methanol, Ricca Chemical
SDP

Catalog No. p-3251193
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 149022-15-1, 67-56-1, 7732-18-5
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00004595
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
IUPAC Name 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.1% (w/v)
CAS Min % 0.12, 86.0, 12.11
CAS Max % 0.13, 89.0, 12.61
Physical Form Liquid
Color Colorless
Solvent or Matrix 90% (v/v) Methanol
Quantity 120 mL
Grade Test Solution
Chemical Name or Material 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.