Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-butanone, 98%

Catalog No. p-3736902
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 563-80-4 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -92.0°C
Color Colorless
Density 0.8000g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3890
Linear Formula (CH3)2CHCOCH3
Quantity 250 mL
Beilstein 01, 682
Specific Gravity 0.8
Merck Index 15, 6162
Solubility Information Solubility in water: 6g/L (20 c). Other solubilities: miscible with most common organic solvents
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-264-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.