Learn More
3-Methyl-2-butanone, 98%
Description
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.
Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | -92°C |
Density | 0.805 |
Boiling Point | 94°C to 95°C |
Flash Point | −1°C (30°F) |
Odor | Fruit-like |
Refractive Index | 1.388 |
Quantity | 1000 mL |
UN Number | UN2397 |
Beilstein | 1071238 |
Merck Index | 14,6090 |
Show More |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 3-METHYLBUTAN-2-ONE |
EINECSNumber | 209-264-3 |
RTECSNumber | EL9100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.