Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-pentanol (mixture of diastereoisomers) 95.0+%, TCI America™
SDP

Catalog No. M085025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M085025ML 25 mL
M08505ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M085025ML Supplier TCI America Supplier No. M085025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 565-60-6
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004528
InChI Key ZXNBBWHRUSXUFZ-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanol, 3-methyl-4-pentanol, 2-pentanol, 3-methyl, 2-hydroxy-3-methylpentane, threo-3-methylpentan-2-ol, 2-pentanol, 3-methyl-, 2s,3r, 3-methyl-pentan-2-ol, acmc-1aozb, acmc-20m2p7, sec-butyl methylcarbinol
PubChem CID 11261
ChEBI CHEBI:77520
IUPAC Name 3-methylpentan-2-ol
SMILES CCC(C)C(C)O

Specifications

Color Colorless
Quantity 25 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pentanol (mixture of diastereoisomers)
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.