Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-3-buten-1-ol, 97%

Catalog No. p-160785
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 763-32-6 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-32-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002933
InChI Key CPJRRXSHAYUTGL-UHFFFAOYSA-N
Synonym 3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
PubChem CID 12988
ChEBI CHEBI:62898
IUPAC Name 3-methylbut-3-en-1-ol
SMILES CC(=C)CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 132°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4327 to 1.4347
Linear Formula H2C=C(CH3)CH2CH2OH
Quantity 100 mL
Specific Gravity 0.85
Solubility Information Solubility in water: 170g/L (20 c)
Formula Weight 86.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-1-ol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-110-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.