Learn More
3-Methyl-3-oxetanemethanol, 97%
Description
3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.
Solubility
Soluble in chloroform, methanol.
Notes
Protect from heat. Store away from oxidizing agents. Store at 4°C.
Specifications
Specifications
| Density | 1.024 |
| Boiling Point | 79°C to 80°C (40 mmHg) |
| Flash Point | 98°C (208°F) |
| Refractive Index | 1.446 |
| Quantity | 100 g |
| Beilstein | 102773 |
| Solubility Information | Soluble in chloroform,methanol. |
| Formula Weight | 102.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Methyl-3-oxetanemethanol |
Safety and Handling
| Hazard Category | H302-H317 |
| Precautionary Statement | P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.