Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-3-pentanol 98.0+%, TCI America™
SDP

Catalog No. p-7132136
Click to view available options
Quantity:
25 mL
100 mL

Chemical Identifiers

CAS 77-74-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004483
InChI Key FRDAATYAJDYRNW-UHFFFAOYSA-N
Synonym Diethylmethylcarbinol
PubChem CID 6493
IUPAC Name 3-methylpentan-3-ol
SMILES CCC(C)(CC)O

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-pentanol
EINECSNumber (2)-0217
RTECSNumber SA7450000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.