Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-3-trimethylsiloxy-1-butyne, 97%

Catalog No. AA4392218
Click to view available options
Quantity:
10 g
50 g

Catalog No. AA4392218

Supplier: Thermo Scientific Chemicals 04392218

Only null left
Add to Cart

CAS: 17869-77-1 | C8H16OSi | 156.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17869-77-1
Molecular Formula C8H16OSi
Molecular Weight (g/mol) 156.3
MDL Number MFCD00053867
InChI Key JNRUXZIXAXHXTN-UHFFFAOYSA-N
Synonym 3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
PubChem CID 87344
IUPAC Name trimethyl(2-methylbut-3-yn-2-yloxy)silane
SMILES CC(C)(C#C)O[Si](C)(C)C

Specifications

Boiling Point 52°C (75 mmHg)
Odor Characteristic
Refractive Index 1.4052
Quantity 50 g
UN Number UN1993
Formula Weight 156.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 241-823-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.