Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-isoxazoleacetic acid, 98%, Thermo Scientific™

Catalog No. AC426650050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19668-85-0
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD01863539
InChI Key POEFJFLAFQWOTL-UHFFFAOYSA-N
Synonym 2-3-methylisoxazol-5-yl acetic acid, 3-methyl-5-isoxazoleacetic acid, 5-isoxazoleacetic acid, 3-methyl, 2-3-methyl-1,2-oxazol-5-yl acetic acid, 3-methyl-1,2-oxazol-5-yl acetic acid, 5-isoxazoleacetic acid,3-methyl, zlchem 1304, 3-methyl-5-isoxazole acetic acid, 3-methyl-isoxazol-5-yl-acetic acid, 5-carboxymethyl-3-methylisoxazole
PubChem CID 4195532
IUPAC Name 2-(3-methyl-1,2-oxazol-5-yl)acetic acid
SMILES CC1=NOC(=C1)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.13
Percent Purity 98%
Physical Form Needle-like Crystals or Fine Crystalline Powder
Chemical Name or Material 3-Methyl-5-isoxazoleacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.