Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-6-nitroindazole 98.0+%, TCI America™
SDP

Catalog No. M235825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M235825G 25 g
M23585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M235825G Supplier TCI America Supplier No. M235825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6494-19-5
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD07082704
InChI Key FUNWSYKLFDLUIZ-UHFFFAOYSA-N
Synonym 3-methyl-6-nitro-1h-indazole, 3-methyl-6-nitroindazole, 1h-indazole, 3-methyl-6-nitro, 3-methyl-6-nitro-indazole, pubchem9492, pubchem18398, 6-nitro-3-methylindazole, ksc352s0d, 2h-indazole,3-methyl-6-nitro
PubChem CID 10921054
IUPAC Name 3-methyl-6-nitro-2H-indazole
SMILES CC1=C2C=CC(=CC2=NN1)[N+](=O)[O-]

Specifications

Melting Point 186°C
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-6-nitroindazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.