Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methylacetophenone, 97%

Catalog No. AC126170250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 585-74-0 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-74-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008742
InChI Key FSPSELPMWGWDRY-UHFFFAOYSA-N
Synonym 3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
PubChem CID 11455
IUPAC Name 1-(3-methylphenyl)ethanone
SMILES CC(=O)C1=CC=CC(C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -9.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 218.0°C to 220.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.529
Linear Formula CH3C6H4COCH3
Quantity 25 g
Beilstein 07,307
Specific Gravity 1.01
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3′-Methylacetophenone
Show More Show Less
Hazard Category Warning
EINECSNumber 209-561-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.