Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylamino-1,2-propanediol, 99%

Catalog No. aal1265314
Click to view available options
Quantity:
25 g
100 g

CAS: 40137-22-2 | C4H11NO2 | 105.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40137-22-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00082732
InChI Key WOMTYMDHLQTCHY-UHFFFAOYSA-N
Synonym 3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
PubChem CID 2734122
IUPAC Name 3-(methylamino)propane-1,2-diol
SMILES CNCC(CO)O

Specifications

Density 1.093
Boiling Point 239°C to 241°C
Flash Point 152°C (305°F)
Refractive Index 1.4745
Quantity 25 g
UN Number UN2735
Beilstein 1733341
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material 3-Methylamino-1,2-propanediol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 254-809-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.