Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylamino)piperidine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. m21715g
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 127294-77-3
Molecular Formula C6H16Cl2N2
Molecular Weight (g/mol) 187.108
MDL Number MFCD00673143
InChI Key VEXIRMPBAHTVNL-UHFFFAOYSA-N
PubChem CID 24820174
IUPAC Name N-methylpiperidin-3-amine;dihydrochloride
SMILES CNC1CCCNC1.Cl.Cl

Specifications

Melting Point 199°C
Color White
Quantity 5 g
Formula Weight 187.11
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Methylamino)piperidine Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.