Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylanisole, 99%

Catalog No. p-7025784
Click to view available options
Quantity:
50 g
250 g

CAS: 100-84-5 | C8H10O | 122.17 g/mol

3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-84-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008395
InChI Key OSIGJGFTADMDOB-UHFFFAOYSA-N
Synonym 3-methylanisole, 3-methoxytoluene, m-methylanisole, m-cresol methyl ether, m-methoxytoluene, benzene, 1-methoxy-3-methyl, anisole, m-methyl, methyl m-tolyl ether, m-cresyl methyl ether, methyl m-cresyl ether
PubChem CID 7530
IUPAC Name 1-methoxy-3-methylbenzene
SMILES COC1=CC=CC(C)=C1

Specifications

Density 0.969
Boiling Point 175°C to 176°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.513
Quantity 50 g
UN Number UN3271
Beilstein 907023
Solubility Information Soluble in alcohol and insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 3-Methylanisole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 202-893-4
RTECSNumber BZ8778000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.